missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (S)-4-Chloro-3-hydroxybutyrate 95.0+%, TCI America™
Supplier: TCI America C171725G
Specifications
| Ethyl (S)-4-Chloro-3-hydroxybutyrate | |
| Yellow | |
| C6H11ClO3 | |
| 25 g | |
| ZAJNMXDBJKCCAT-YFKPBYRVSA-N | |
| ethyl (3S)-4-chloro-3-hydroxybutanoate | |
| 7019277 | |
| ≥95.0% (GC) |
| 86728-85-0 | |
| 96°C | |
| MFCD00211241 | |
| ethyl s-4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutanoate, ethyl 3s-4-chloro-3-hydroxybutanoate, ethyl s-4-chloro-3-hydroxybutanoate, s-4-chloro-3-hydroxybutyrate, s-4-chloro-3-hydroxybutyric acid ethyl ester, s-4-chloro-3-hydroxy-n-butyric acid ethyl ester, s-ethyl 4-chloro-3-hydroxybutanoate | |
| CCOC(=O)C[C@H](O)CCl | |
| 166.60 | |
| 166.60 | |
| Liquid |
Chemical Identifiers
| 86728-85-0 | |
| 166.60 | |
| ZAJNMXDBJKCCAT-YFKPBYRVSA-N | |
| 7019277 | |
| CCOC(=O)C[C@H](O)CCl |
| C6H11ClO3 | |
| MFCD00211241 | |
| ethyl s-4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutanoate, ethyl 3s-4-chloro-3-hydroxybutanoate, ethyl s-4-chloro-3-hydroxybutanoate, s-4-chloro-3-hydroxybutyrate, s-4-chloro-3-hydroxybutyric acid ethyl ester, s-4-chloro-3-hydroxy-n-butyric acid ethyl ester, s-ethyl 4-chloro-3-hydroxybutanoate | |
| ethyl (3S)-4-chloro-3-hydroxybutanoate |
Safety and Handling
EINECSNumber : (2)-3782&(2)-3796&(2)-3882
TSCA : No
Recommended Storage : Refrigerator