missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate, 98%

CAS: 86728-85-0 | C6H11ClO3 | 166.60 g/mol

$29.30 - $97.45

Chemical Identifiers

CAS 86728-85-0
Molecular Formula C6H11ClO3
Molecular Weight (g/mol) 166.60
MDL Number MFCD00211241
InChI Key ZAJNMXDBJKCCAT-YFKPBYRVSA-N
Synonym ethyl s-4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutanoate, ethyl 3s-4-chloro-3-hydroxybutanoate, ethyl s-4-chloro-3-hydroxybutanoate, s-4-chloro-3-hydroxybutyrate, s-4-chloro-3-hydroxybutyric acid ethyl ester, s-4-chloro-3-hydroxy-n-butyric acid ethyl ester, s-ethyl 4-chloro-3-hydroxybutanoate
PubChem CID 7019277
IUPAC Name ethyl (3S)-4-chloro-3-hydroxybutanoate
SMILES CCOC(=O)C[C@H](O)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1855403
View Documents
Thermo Scientific Chemicals
L1855403
1 g
Each for $29.30
Only null left
 
AAL1855406
View Documents
Thermo Scientific Chemicals
L1855406
5 g
Each for $97.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

86728-85-0
166.60
ZAJNMXDBJKCCAT-YFKPBYRVSA-N
7019277
CCOC(=O)C[C@H](O)CCl
C6H11ClO3
MFCD00211241
ethyl s-4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutanoate, ethyl 3s-4-chloro-3-hydroxybutanoate, ethyl s-4-chloro-3-hydroxybutanoate, s-4-chloro-3-hydroxybutyrate, s-4-chloro-3-hydroxybutyric acid ethyl ester, s-4-chloro-3-hydroxy-n-butyric acid ethyl ester, s-ethyl 4-chloro-3-hydroxybutanoate
ethyl (3S)-4-chloro-3-hydroxybutanoate

Specifications

86728-85-0
93°C to 95°C (5 mmHg)
C6H11ClO3
MFCD00211241
ethyl s-4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutanoate, ethyl 3s-4-chloro-3-hydroxybutanoate, ethyl s-4-chloro-3-hydroxybutanoate, s-4-chloro-3-hydroxybutyrate, s-4-chloro-3-hydroxybutyric acid ethyl ester, s-4-chloro-3-hydroxy-n-butyric acid ethyl ester, s-ethyl 4-chloro-3-hydroxybutanoate
−14° (Neat)
ethyl (3S)-4-chloro-3-hydroxybutanoate
7019277
98%
ee 98%
1.19
>110°C (230°F)
1.453
1 g
ZAJNMXDBJKCCAT-YFKPBYRVSA-N
CCOC(=O)C[C@H](O)CCl
166.60
166.61
Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H312-H332-H315
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.

P280-P305+P351+P338-P310

H318

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only