Learn More
Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate, 98%
CAS: 86728-85-0 | C6H11ClO3 | 166.60 g/mol
Supplier: Thermo Scientific Chemicals L1855406
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate | |
| 1.19 | |
| >110°C (230°F) | |
| C6H11ClO3 | |
| MFCD00211241 | |
| ethyl s-4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutanoate, ethyl 3s-4-chloro-3-hydroxybutanoate, ethyl s-4-chloro-3-hydroxybutanoate, s-4-chloro-3-hydroxybutyrate, s-4-chloro-3-hydroxybutyric acid ethyl ester, s-4-chloro-3-hydroxy-n-butyric acid ethyl ester, s-ethyl 4-chloro-3-hydroxybutanoate | |
| −14° (Neat) | |
| ethyl (3S)-4-chloro-3-hydroxybutanoate | |
| 7019277 | |
| 98% |
| 86728-85-0 | |
| 93°C to 95°C (5 mmHg) | |
| ee 98% | |
| 1.453 | |
| 5 g | |
| ZAJNMXDBJKCCAT-YFKPBYRVSA-N | |
| CCOC(=O)C[C@H](O)CCl | |
| 166.60 | |
| 166.61 |
Chemical Identifiers
| 86728-85-0 | |
| 166.60 | |
| ZAJNMXDBJKCCAT-YFKPBYRVSA-N | |
| 7019277 | |
| CCOC(=O)C[C@H](O)CCl |
| C6H11ClO3 | |
| MFCD00211241 | |
| ethyl s-4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutanoate, ethyl 3s-4-chloro-3-hydroxybutanoate, ethyl s-4-chloro-3-hydroxybutanoate, s-4-chloro-3-hydroxybutyrate, s-4-chloro-3-hydroxybutyric acid ethyl ester, s-4-chloro-3-hydroxy-n-butyric acid ethyl ester, s-ethyl 4-chloro-3-hydroxybutanoate | |
| ethyl (3S)-4-chloro-3-hydroxybutanoate |
Safety and Handling
GHS H Statement
H318-H302-H312-H332-H315
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
P280-P305+P351+P338-P310
H318
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only