missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-(-)-1-Ethyl-2-pyrrolidinecarboxylate 98.0+%, TCI America™

Catalog No. E04505G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04505G 5 g
E04501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04505G Supplier TCI America Supplier No. E04505G

Chemical Identifiers

CAS 938-54-5
Molecular Formula C9H17NO2
Molecular Weight (g/mol) 171.24
MDL Number MFCD00191434
InChI Key CUDOPASYFARLIF-QMMMGPOBSA-N
Synonym (S)-(-)-1-Ethyl-2-pyrrolidinecarboxylic Acid Ethyl Ester, (-)-1-Ethyl-L-proline Ethyl Ester, Et-Pro-OEt
PubChem CID 10942897
IUPAC Name ethyl (2S)-1-ethylpyrrolidine-2-carboxylate
SMILES CCN1CCCC1C(=O)OCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 171.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl (S)-(-)-1-Ethyl-2-pyrrolidinecarboxylate