accessibility menu, dialog, popup

UserName

CH3NOOH3C

CAS RN 938-54-5

IUPAC Name: ethyl 1-ethylpyrrolidine-2-carboxylate
Synonyms: ethyl 1-ethylpyrrolidine-2-carboxylate
Molecular Weight (g/mol): 171.24
Molecular Formula: C9H17NO2
InChi Key: CUDOPASYFARLIF-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCCN1CC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results