missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%, Thermo Scientific Chemicals

Catalog No. p-7041508
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH6237803 1 g
AAH6237806 5 g
2 options

Catalog No. AAH6237803

Supplier: Thermo Scientific Chemicals H6237803

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144978-12-1
Molecular Formula C12H19NO5
Molecular Weight (g/mol) 257.29
MDL Number MFCD07778090
InChI Key YWWWGFSJHCFVOW-UHFFFAOYNA-N
Synonym boc-pyr-oet, s-ethyl-n-boc-pyroglutamate, n-boc-l-pyroglutamic acid ethyl ester, 1-boc-l-pyroglutamic acid ethyl ester, boc-l-pyroglutamic acid ethyl ester, s-1-tert-butyl 2-ethyl 5-oxopyrrolidine-1,2-dicarboxylate, ethyl n-boc-l-pyroglutamate, 1-tert-butyl 2-ethyl 2s-5-oxopyrrolidine-1,2-dicarboxylate, s-n-alpha-t-butyloxycarbonyl-pyroglutamic acid ethyl ester, 1-tert-butyl 2-ethyl s-5-oxopyrrolidine-1,2-dicarboxylate
PubChem CID 5461186
SMILES CCOC(=O)C1CCC(=O)N1C(=O)OC(C)(C)C

Specifications

Quantity 1 g
IUPAC Name 1-tert-butyl 2-ethyl 5-oxopyrrolidine-1,2-dicarboxylate
Formula Weight 257.28
Percent Purity 98%
Chemical Name or Material Ethyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only