missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-(-)-1-Boc-4-oxopiperidine-2-carboxylate, 95%, Thermo Scientific Chemicals

Catalog No. AAH5721303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5721303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5721303 Supplier Thermo Scientific Chemicals Supplier No. H5721303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

1-Boc-4-oxopiperidine-2-carboxylate participates in the one-pot in situ formation and reaction of trimethyl (trichloromethyl) silane which finds application in the synthesis of 2, 2, 2-trichloromethylcarbinols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-4-oxopiperidine-2-carboxylate participates in the one-pot in situ formation and reaction of trimethyl (trichloromethyl) silane which finds application in the synthesis of 2, 2, 2-trichloromethylcarbinols.

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.

Chemical Identifiers

CAS 180854-44-8
Molecular Formula C13H21NO5
Molecular Weight (g/mol) 271.313
MDL Number MFCD15071881
InChI Key YAVQLRUBKDCOCI-JTQLQIEISA-N
Synonym ethyl s-1-boc-4-oxopiperidine-2-carboxylate, s-1-tert-butyl 2-ethyl 4-oxopiperidine-1,2-dicarboxylate, 1-tert-butyl 2-ethyl 2s-4-oxopiperidine-1,2-dicarboxylate, ethyl r-+-1-boc-4-oxopiperidine-2-carboxylate, ethyl s---1-boc-4-oxopiperidine-2-carboxylate, ethyl s-n-tert-butoxycarbonyl-4-oxo-2-pipecolate, o1-tert-butyl o2-ethyl 2s-4-oxopiperidine-1,2-dicarboxylate, 2s-4-oxopiperidine-1,2-dicarboxylic acid 1-tert-butyl 2-ethyl ester
PubChem CID 11369276
IUPAC Name 1-O-tert-butyl 2-O-ethyl (2S)-4-oxopiperidine-1,2-dicarboxylate
SMILES CCOC(=O)C1CC(=O)CCN1C(=O)OC(C)(C)C

Specifications

Density 1.079
Flash Point >110°C (230°F)
Refractive Index 1.46
Quantity 1 g
Optical Rotation −20° (c=0.5 in Chloroform)
Formula Weight 271.31
Percent Purity 95%
Chemical Name or Material Ethyl (S)-(-)-1-Boc-4-oxopiperidine-2-carboxylate
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only