accessibility menu, dialog, popup

UserName

CAS RN 180854-44-8

CH3OOONOOCH3H3CCH3

CAS RN 180854-44-8

IUPAC Name: 1-tert-butyl 2-ethyl 4-oxopiperidine-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-ethyl 4-oxopiperidine-1,2-dicarboxylate
Molecular Weight (g/mol): 271.31
Molecular Formula: C13H21NO5
InChi Key: YAVQLRUBKDCOCI-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CC(=O)CCN1C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results