missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (R)-1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate 98.0+%, TCI America™

Catalog No. E09485G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E09485G Supplier TCI America Supplier No. E09485G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 194726-40-4
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.33
MDL Number MFCD07374389
InChI Key YCXCRFGBFZTUSU-SNVBAGLBSA-N
Synonym r-1-tert-butyl 3-ethyl piperidine-1,3-dicarboxylate, ethyl r-1-boc-nipecotate, r-1-boc-piperidine-3-carboxylic acid ethyl ester, ethyl r-n-boc-piperidine-3-carboxylate, ethyl 1-boc-d-nipecotate, r-ethyl n-boc-piperidine-3-carboxylate, r---n-boc-nipecotic acid ethyl ester, ethyl 3r-1-tert-butoxycarbonyl-3-piperidinecarboxylate, ethyl r-1-boc-3-piperidinecarboxylate, r-1-boc-3-piperidinecarboxylate ethyl ester
PubChem CID 1514446
IUPAC Name 1-O-tert-butyl 3-O-ethyl (3R)-piperidine-1,3-dicarboxylate
SMILES CCOC(=O)C1CCCN(C1)C(=O)OC(C)(C)C

Specifications

Melting Point 37°C
Color White-Yellow
Quantity 5 g
Formula Weight 257.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl (R)-1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate