accessibility menu, dialog, popup

UserName

CAS RN 194726-40-4

IUPAC Name: 1-tert-butyl 3-ethyl piperidine-1,3-dicarboxylate
Synonyms: 1-tert-butyl 3-ethyl piperidine-1,3-dicarboxylate
Molecular Weight (g/mol): 257.33
Molecular Formula: C13H23NO4
InChi Key: YCXCRFGBFZTUSU-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCCN(C1)C(=O)OC(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results