missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Pyruvate 97.0+%, TCI America™

Catalog No. P089125ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P089125ML 25 mL
P0891100ML 100 mL
P0891500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P089125ML Supplier TCI America Supplier No. P089125ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 617-35-6
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00009123
InChI Key XXRCUYVCPSWGCC-UHFFFAOYSA-N
Synonym ethyl pyruvate, ethyl 2-oxopropionate, pyruvic acid, ethyl ester, propanoic acid, 2-oxo-, ethyl ester, pyruvic acid ethyl ester, ethyl pyroracemate, ethyl acetylformate, 2-oxo-propionic acid ethyl ester, ethyl pyruvate natural, ethyl alpha-ketopropionate
PubChem CID 12041
IUPAC Name ethyl 2-oxopropanoate
SMILES CCOC(=O)C(=O)C

Specifications

Melting Point -50°C
Color Yellow
Boiling Point 142°C
Quantity 25 mL
UN Number 3272
Formula Weight 116.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Pyruvate
EINECSNumber (2)-1505
TSCA Yes