accessibility menu, dialog, popup

UserName

H3COOCH3O

CAS RN 617-35-6

IUPAC Name: ethyl 2-oxopropanoate
Synonyms: ethyl pyruvate
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: XXRCUYVCPSWGCC-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)=O
Boiling Point 
  • (3)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (6)
  • (1)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (3)
  • (1)
  • (6)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)