Learn More
Ethyl pyruvate, 98%
CAS: 617-35-6 | C5H8O3 | 116.12 g/mol
$181.02 - $181.02
Chemical Identifiers
| CAS | 617-35-6 |
|---|---|
| Molecular Formula | C5H8O3 |
| Molecular Weight (g/mol) | 116.12 |
| MDL Number | MFCD00009123 |
| InChI Key | XXRCUYVCPSWGCC-UHFFFAOYSA-N |
| Synonym | ethyl pyruvate, ethyl 2-oxopropionate, pyruvic acid, ethyl ester, propanoic acid, 2-oxo-, ethyl ester, pyruvic acid ethyl ester, ethyl pyroracemate, ethyl acetylformate, 2-oxo-propionic acid ethyl ester, ethyl pyruvate natural, ethyl alpha-ketopropionate |
| PubChem CID | 12041 |
| IUPAC Name | ethyl 2-oxopropanoate |
| SMILES | CCOC(=O)C(=O)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC118941000
|
Thermo Scientific Chemicals
118941000 |
100 g | Glass bottle |
Each for $181.02
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 617-35-6 | |
| 116.12 | |
| XXRCUYVCPSWGCC-UHFFFAOYSA-N | |
| 12041 | |
| CCOC(=O)C(=O)C |
| C5H8O3 | |
| MFCD00009123 | |
| ethyl pyruvate, ethyl 2-oxopropionate, pyruvic acid, ethyl ester, propanoic acid, 2-oxo-, ethyl ester, pyruvic acid ethyl ester, ethyl pyroracemate, ethyl acetylformate, 2-oxo-propionic acid ethyl ester, ethyl pyruvate natural, ethyl alpha-ketopropionate | |
| ethyl 2-oxopropanoate |
Specifications
| 617-35-6 | |
| Yellow | |
| 148.0°C to 150.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4040 to 1.4060 | |
| MFCD00009123 | |
| 03, 616 | |
| 15, 8135 | |
| Solubility in water: 10g/L (20°C). Other solubilities: soluble in ethanol,ether | |
| CCOC(=O)C(=O)C | |
| 116.12 | |
| 1.18 mPa.s | |
| 98% | |
| Ethyl pyruvate, 98% |
| -58.0°C | |
| 1.0453g/mL | |
| 45°C | |
| 98% | |
| C5H8O3 | |
| CH3COCO2C2H5 | |
| 100 g | |
| 1.0453 | |
| ethyl pyruvate, ethyl 2-oxopropionate, pyruvic acid, ethyl ester, propanoic acid, 2-oxo-, ethyl ester, pyruvic acid ethyl ester, ethyl pyroracemate, ethyl acetylformate, 2-oxo-propionic acid ethyl ester, ethyl pyruvate natural, ethyl alpha-ketopropionate | |
| XXRCUYVCPSWGCC-UHFFFAOYSA-N | |
| ethyl 2-oxopropanoate | |
| 12041 | |
| 116.12 | |
| Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep contai
GHS Signal Word: Warning
EINECSNumber : 210-511-2
TSCA : TSCA
RUO – Research Use Only