missing translation for 'onlineSavingsMsg'
Learn More

Ethyl pyruvate, 98%

CAS: 617-35-6 | C5H8O3 | 116.116 g/mol

Supplier:  Thermo Scientific Chemicals A1504530

 View more versions of this product

Catalog No. AAA1504530


Only null left
Explore available promotions

Description

Description

Ethyl pyruvate is used as a flavoring agent in food and anti-inflammatory agent for the treatment of critical inflammatory conditions. It is used in the treatment of critical illnesses such as severe sepsis, acute respiratory distress syndrome, burn injury, acute pancreatitis and stroke. It plays an important role in cardiac function after coronary ischemia and reperfusion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with water, ethanol and ether.

Notes
Store in a cool place. Incompatible with oxidizing agents.
Specifications

Specifications

Ethyl pyruvate
-58°C
143°C to 145°C
98%
C5H8O3
MFCD00009123
UN3272
14,8021
Miscible with water,ethanol and ether.
CCOC(=O)C(=O)C
116.116
116.12
617-35-6
1.05
45°C (113°F)
Pleasant
1.405
250 g
1071466
ethyl pyruvate, ethyl 2-oxopropionate, pyruvic acid, ethyl ester, propanoic acid, 2-oxo-, ethyl ester, pyruvic acid ethyl ester, ethyl pyroracemate, ethyl acetylformate, 2-oxo-propionic acid ethyl ester, ethyl pyruvate natural, ethyl alpha-ketopropionate
XXRCUYVCPSWGCC-UHFFFAOYSA-N
ethyl 2-oxopropanoate
12041
98%

Chemical Identifiers

617-35-6
116.116
XXRCUYVCPSWGCC-UHFFFAOYSA-N
12041
CCOC(=O)C(=O)C
C5H8O3
MFCD00009123
ethyl pyruvate, ethyl 2-oxopropionate, pyruvic acid, ethyl ester, propanoic acid, 2-oxo-, ethyl ester, pyruvic acid ethyl ester, ethyl pyroracemate, ethyl acetylformate, 2-oxo-propionic acid ethyl ester, ethyl pyruvate natural, ethyl alpha-ketopropionate
ethyl 2-oxopropanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 210-511-2

TSCA : Yes

storageNote1 : Store cold

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only