missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Pyrrole-2-carboxylate 98.0+%, TCI America™
Supplier: TCI America E103025G
Specifications
| Ethyl Pyrrole-2-carboxylate | |
| 41°C | |
| C7H9NO2 | |
| 25 g | |
| PAEYAKGINDQUCT-UHFFFAOYSA-N | |
| ethyl 1H-pyrrole-2-carboxylate | |
| 255670 | |
| ≥98.0% (GC) |
| 2199-43-1 | |
| 235°C | |
| MFCD00817049 | |
| ethyl pyrrole-2-carboxylate, ethyl 2-pyrrolecarboxylate, ethyl pyrrole-2-carbonate, ethylpyrrole-2-carboxylate, 1h-pyrrole-2-carboxylic acid, ethyl ester, 2-ethoxycarbonyl-1h-pyrrole, pyrrole-2-carboxylic acid, ethyl ester, 1h-pyrrole-2-carboxylic acid ethyl ester, pyrrole-2-carboxylic acid ethyl ester, ethyl1h-pyrrole-2-carboxylate | |
| CCOC(=O)C1=CC=CN1 | |
| 139.154 | |
| 139.15 | |
| Crystalline Powder |
Chemical Identifiers
| 2199-43-1 | |
| 139.154 | |
| PAEYAKGINDQUCT-UHFFFAOYSA-N | |
| 255670 | |
| CCOC(=O)C1=CC=CN1 |
| C7H9NO2 | |
| MFCD00817049 | |
| ethyl pyrrole-2-carboxylate, ethyl 2-pyrrolecarboxylate, ethyl pyrrole-2-carbonate, ethylpyrrole-2-carboxylate, 1h-pyrrole-2-carboxylic acid, ethyl ester, 2-ethoxycarbonyl-1h-pyrrole, pyrrole-2-carboxylic acid, ethyl ester, 1h-pyrrole-2-carboxylic acid ethyl ester, pyrrole-2-carboxylic acid ethyl ester, ethyl1h-pyrrole-2-carboxylate | |
| ethyl 1H-pyrrole-2-carboxylate |
Safety and Handling
TSCA : No