missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Pyrimidine-4-carboxylate 98.0+%, TCI America™
Supplier: TCI America E088125G
Specifications
| Ethyl Pyrimidine-4-carboxylate | |
| 39°C | |
| 160°C | |
| MFCD09832999 | |
| ethyl 4-pyrimidinecarboxylate, ethyl4-pyrimidinecarboxylate, pyrimidine-4-carboxylic acid ethyl ester, 4-pyrimidinecarboxylic acid, ethyl ester, pubchem20807, 4-ethoxycarbonylpyrimidine, ethylpyrimidine-4-carboxylate, 2-ethylpyrimidine-4-carboxylate, 4-pyrimidinecarboxylicacid, ethyl ester, pyrimidine-6-carboxylic acid ethyl ester | |
| CCOC(=O)C1=NC=NC=C1 | |
| 152.153 | |
| 152.15 | |
| Crystalline Lumps |
| 62846-82-6 | |
| White-Yellow | |
| C7H8N2O2 | |
| 25 g | |
| DWRWSNAREGLUHZ-UHFFFAOYSA-N | |
| ethyl pyrimidine-4-carboxylate | |
| 13639438 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 62846-82-6 | |
| 152.153 | |
| DWRWSNAREGLUHZ-UHFFFAOYSA-N | |
| 13639438 | |
| CCOC(=O)C1=NC=NC=C1 |
| C7H8N2O2 | |
| MFCD09832999 | |
| ethyl 4-pyrimidinecarboxylate, ethyl4-pyrimidinecarboxylate, pyrimidine-4-carboxylic acid ethyl ester, 4-pyrimidinecarboxylic acid, ethyl ester, pubchem20807, 4-ethoxycarbonylpyrimidine, ethylpyrimidine-4-carboxylate, 2-ethylpyrimidine-4-carboxylate, 4-pyrimidinecarboxylicacid, ethyl ester, pyrimidine-6-carboxylic acid ethyl ester | |
| ethyl pyrimidine-4-carboxylate |
Safety and Handling
TSCA : No