missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Propyl Sulfide 96.0+%, TCI America™
Supplier: TCI America E022410G
Specifications
| Ethyl Propyl Sulfide | |
| Colorless | |
| C5H12S | |
| 10 g | |
| ZDDDFDQTSXYYSE-UHFFFAOYSA-N | |
| 1-ethylsulfanylpropane | |
| 20063 | |
| ≥96.0% (GC) |
| 4110-50-3 | |
| 117°C | |
| MFCD00039940 | |
| 1993 | |
| CCCSCC | |
| 104.211 | |
| 104.21 | |
| Liquid |
Chemical Identifiers
| 4110-50-3 | |
| 104.211 | |
| ZDDDFDQTSXYYSE-UHFFFAOYSA-N | |
| 1-ethylsulfanylpropane |
| C5H12S | |
| MFCD00039940 | |
| 20063 | |
| CCCSCC |
Safety and Handling
TSCA : No