missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Propyl Ether 98.0+%, TCI America™

Catalog No. E005225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E005225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E005225ML Supplier TCI America Supplier No. E005225ML
Only null left

Chemical Identifiers

CAS 628-32-0
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00015215
InChI Key NVJUHMXYKCUMQA-UHFFFAOYSA-N
Synonym 3-Ethoxypropane
PubChem CID 12340
IUPAC Name 1-ethoxypropane
SMILES CCCOCC

Specifications

Color Colorless
Boiling Point 62°C
Quantity 25 mL
UN Number 2615
Formula Weight 88.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Propyl Ether
RTECSNumber KO0600000
TSCA No
Recommended Storage Refrigerator