CAS RN 628-32-0
IUPAC Name:
1-ethoxypropane
Synonyms:
ethyl propyl ether
Molecular Weight (g/mol):
88.15
Molecular Formula:
C5H12O
InChi Key:
NVJUHMXYKCUMQA-UHFFFAOYSA-N
SMILES:
CCCOCC
1
–
1
of
1
results
1
Ethyl Propyl Ether 98.0+%, TCI America™
CAS: 628-32-0 Molecular Formula: C5H12O Molecular Weight (g/mol): 88.15 MDL Number: MFCD00015215 InChI Key: NVJUHMXYKCUMQA-UHFFFAOYSA-N Synonym: 3-Ethoxypropane PubChem CID: 12340 IUPAC Name: 1-ethoxypropane SMILES: CCCOCC