Learn More
Ethyl propionylacetate, 95%
CAS: 4949-44-4 | C7H12O3 | 144.17 g/mol
Supplier: Thermo Scientific Chemicals A1546414
Description
Ethyl propionylacetate acts as a precursor used in the preparation of 1-ethyl- and 1-n-propyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid bearing one or two substituents on the benzene ring. Further, it is used to prepare 3-propionyl-2H-cyclohepta[b]furan-2-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl propionylacetate acts as a precursor used in the preparation of 1-ethyl- and 1-n-propyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid bearing one or two substituents on the benzene ring. Further, it is used to prepare 3-propionyl-2H-cyclohepta[b]furan-2-one.
Solubility
Miscible with glacial acetic acid.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
Specifications
| Ethyl propionylacetate | |
| 1.012 | |
| 77°C (170°F) | |
| 1.422 | |
| 25 g | |
| ethyl propionylacetate, ethyl 3-oxovalerate, ethyl 3-oxo-n-valerate, pentanoic acid, 3-oxo-, ethyl ester, 3-oxopentanoic acid ethyl ester, 3-oxo-pentanoic acid ethyl ester, ethyl 3-ketopentanoate, 3-oxovaleric acid ethyl ester, propionylacetic acid ethyl ester, ethylpropanylacetate | |
| UDRCONFHWYGWFI-UHFFFAOYSA-N | |
| ethyl 3-oxopentanoate | |
| 78656 | |
| 95% |
| 4949-44-4 | |
| 190°C to 192°C | |
| C7H12O3 | |
| MFCD00009317 | |
| 970270 | |
| Miscible with glacial acetic acid. | |
| CCC(=O)CC(=O)OCC | |
| 144.17 | |
| 144.17 |
Chemical Identifiers
| 4949-44-4 | |
| 144.17 | |
| UDRCONFHWYGWFI-UHFFFAOYSA-N | |
| 78656 | |
| CCC(=O)CC(=O)OCC |
| C7H12O3 | |
| MFCD00009317 | |
| ethyl propionylacetate, ethyl 3-oxovalerate, ethyl 3-oxo-n-valerate, pentanoic acid, 3-oxo-, ethyl ester, 3-oxopentanoic acid ethyl ester, 3-oxo-pentanoic acid ethyl ester, ethyl 3-ketopentanoate, 3-oxovaleric acid ethyl ester, propionylacetic acid ethyl ester, ethylpropanylacetate | |
| ethyl 3-oxopentanoate |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 225-593-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only