missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl propionylacetate, 95%
Description
Ethyl propionylacetate acts as a precursor used in the preparation of 1-ethyl- and 1-n-propyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid bearing one or two substituents on the benzene ring. Further, it is used to prepare 3-propionyl-2H-cyclohepta[b]furan-2-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl propionylacetate acts as a precursor used in the preparation of 1-ethyl- and 1-n-propyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid bearing one or two substituents on the benzene ring. Further, it is used to prepare 3-propionyl-2H-cyclohepta[b]furan-2-one.
Solubility
Miscible with glacial acetic acid.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
Ethyl propionylacetate acts as a precursor used in the preparation of 1-ethyl- and 1-n-propyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid bearing one or two substituents on the benzene ring. Further, it is used to prepare 3-propionyl-2H-cyclohepta[b]furan-2-one.
Solubility
Miscible with glacial acetic acid.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
| Density | 1.012 |
| Boiling Point | 190°C to 192°C |
| Flash Point | 77°C (170°F) |
| Refractive Index | 1.422 |
| Quantity | 5 g |
| Beilstein | 970270 |
| Solubility Information | Miscible with glacial acetic acid. |
| Formula Weight | 144.17 |
| Percent Purity | 95% |
| Chemical Name or Material | Ethyl propionylacetate |
Safety and Handling
| Hazard Category | H227-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
| Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| EINECSNumber | 225-593-5 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only