missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Propiolate 98.0+%, TCI America™

Catalog No. p-7134663
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
5 mL
Catalog No. Quantity
P05295ML 5 mL
P052925ML 25 mL
2 options

Catalog No. P05295ML

Supplier: TCI America P05295ML

Only null left

Chemical Identifiers

CAS 623-47-2
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00009184
InChI Key FMVJYQGSRWVMQV-UHFFFAOYSA-N
Synonym ethyl propiolate, ethyl acetylenecarboxylate, 2-propynoic acid, ethyl ester, propiolic acid ethyl ester, ethyl propynoate, ethoxycarbonyl acetylene, ethyl 2-propynoate, propiolic acid, ethyl ester, unii-w235g5u52s, propynoic acid ethyl ester
PubChem CID 12182
ChEBI CHEBI:51740
IUPAC Name ethyl prop-2-ynoate
SMILES CCOC(=O)C#C

Specifications

Melting Point 9°C
Color Yellow
Boiling Point 119°C
Quantity 5 mL
UN Number 3272
Formula Weight 98.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Propiolate