accessibility menu, dialog, popup

UserName

H3COOCH

CAS RN 623-47-2

IUPAC Name: ethyl prop-2-ynoate
Synonyms: ethyl propiolate
Molecular Weight (g/mol): 98.1
Molecular Formula: C5H6O2
InChi Key: FMVJYQGSRWVMQV-UHFFFAOYSA-N
SMILES: CCOC(=O)C#C
Boiling Point 
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (6)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (5)