missing translation for 'onlineSavingsMsg'
Learn More

Ethyl propiolate, 99%

Catalog No. p-7022343
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1506806 5 g
AAA1506814 25 g
AAA1506822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1506806 Supplier Thermo Scientific Chemicals Supplier No. A1506806
Only null left

CAS: 623-47-2 | C5H6O2 | 98.10 g/mol

Ethyl propiolate is used as a precursor in the preparation of substituted anthraquinones and pyrazolo[1,5-a]pyridine, which exhibits anti-inflammatory activity. Further, it is employed in the synthesis of benzene-1,2,3,5-tetracarboxylates promoted by triphenylphosphine. Its halogenated derivatives are used as a substrate for direct, asymmetric alkynylation of cyclic beta-ketoesters using chiral phase-transfer catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl propiolate is used as a precursor in the preparation of substituted anthraquinones and pyrazolo[1,5-a]pyridine, which exhibits anti-inflammatory activity. Further, it is employed in the synthesis of benzene-1,2,3,5-tetracarboxylates promoted by triphenylphosphine. Its halogenated derivatives are used as a substrate for direct, asymmetric alkynylation of cyclic beta-ketoesters using chiral phase-transfer catalysts.

Solubility
Miscible with alcohol.

Notes
Incompatible with oxidizing agents, bases and acids.

Chemical Identifiers

CAS 623-47-2
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00009184
InChI Key FMVJYQGSRWVMQV-UHFFFAOYSA-N
Synonym ethyl propiolate, ethyl acetylenecarboxylate, 2-propynoic acid, ethyl ester, propiolic acid ethyl ester, ethyl propynoate, ethoxycarbonyl acetylene, ethyl 2-propynoate, propiolic acid, ethyl ester, unii-w235g5u52s, propynoic acid ethyl ester
PubChem CID 12182
ChEBI CHEBI:51740
IUPAC Name ethyl prop-2-ynoate
SMILES CCOC(=O)C#C

Specifications

Melting Point 9°C
Density 0.963
Boiling Point 118°C to 120°C
Flash Point 23°C (73°F)
Odor Pungent
Refractive Index 1.412
Linear Formula HC≡CCO2CH2CH3
Quantity 5 g
UN Number UN3272
Beilstein 878250
Solubility Information Miscible with alcohol.
Formula Weight 98.1
Percent Purity 99%
Chemical Name or Material Ethyl propiolate
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 210-795-8
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only