Learn More
Ethyl pipecolinate, 98+%
CAS: 15862-72-3 | C8H16NO2 | 158.22 g/mol
$52.44 - $457.22
Chemical Identifiers
| CAS | 15862-72-3 |
|---|---|
| Molecular Formula | C8H16NO2 |
| Molecular Weight (g/mol) | 158.22 |
| MDL Number | MFCD00005980 |
| InChI Key | SZIKRGHFZTYTIT-ZETCQYMHSA-O |
| Synonym | ethyl pipecolinate, ethyl 2-piperidinecarboxylate, ethyl pipecolate, pipecolinic acid ethyl ester, pipecolic acid ethyl ester, ethylpiperidine-2-carboxylate, 2-piperidinecarboxylic acid ethyl ester, 2-piperidinecarboxylic acid, ethyl ester, ethylpipecolinate, pubchem7720 |
| PubChem CID | 27517 |
| IUPAC Name | ethyl piperidine-2-carboxylate |
| SMILES | CCOC(=O)[C@@H]1CCCC[NH2+]1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1053203
|
Thermo Scientific Chemicals
L1053203 |
1 g |
Each for $52.44
|
|
|||||
|
AAL1053206
|
Thermo Scientific Chemicals
L1053206 |
5 g |
Each for $149.70
|
|
|||||
|
AAL1053214
|
Thermo Scientific Chemicals
L1053214 |
25 g |
Each for $457.22
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 15862-72-3 | |
| 158.22 | |
| SZIKRGHFZTYTIT-ZETCQYMHSA-O | |
| 27517 | |
| CCOC(=O)[C@@H]1CCCC[NH2+]1 |
| C8H16NO2 | |
| MFCD00005980 | |
| ethyl pipecolinate, ethyl 2-piperidinecarboxylate, ethyl pipecolate, pipecolinic acid ethyl ester, pipecolic acid ethyl ester, ethylpiperidine-2-carboxylate, 2-piperidinecarboxylic acid ethyl ester, 2-piperidinecarboxylic acid, ethyl ester, ethylpipecolinate, pubchem7720 | |
| ethyl piperidine-2-carboxylate |
Specifications
| 15862-72-3 | |
| 216°C to 217°C | |
| C8H16NO2 | |
| MFCD00005980 | |
| UN1993 | |
| ethyl pipecolinate, ethyl 2-piperidinecarboxylate, ethyl pipecolate, pipecolinic acid ethyl ester, pipecolic acid ethyl ester, ethylpiperidine-2-carboxylate, 2-piperidinecarboxylic acid ethyl ester, 2-piperidinecarboxylic acid, ethyl ester, ethylpipecolinate, pubchem7720 | |
| CCOC(=O)[C@@H]1CCCC[NH2+]1 | |
| 158.22 | |
| 157.21 | |
| Ethyl pipecolinate |
| 1.006 | |
| 46°C (114°F) | |
| 1.456 | |
| 1 g | |
| 81694 | |
| SZIKRGHFZTYTIT-ZETCQYMHSA-O | |
| ethyl piperidine-2-carboxylate | |
| 27517 | |
| ≥98% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 239-990-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only