accessibility menu, dialog, popup

UserName

CAS RN 15862-72-3

H3COO(S)H2N

CAS RN 15862-72-3

IUPAC Name: (2S)-2-(ethoxycarbonyl)piperidin-1-ium
Synonyms: (2S)-2-(ethoxycarbonyl)piperidin-1-ium
Molecular Weight (g/mol): 158.22
Molecular Formula: C8H16NO2
InChi Key: SZIKRGHFZTYTIT-ZETCQYMHSA-O
SMILES: CCOC(=O)[C@@H]1CCCC[NH2+]1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)