missing translation for 'onlineSavingsMsg'
Learn More

Ethyl picolinate, 99%

CAS: 2524-52-9 | C8H9NO2 | 151.17 g/mol

Supplier:  Thermo Scientific Chemicals A1397322

 View more versions of this product

Catalog No. AAA1397322


Only null left
Explore available promotions

Description

Description

Ethyl picolinate is used in the preparation of 2-Aminodihydro[1,3]thiazines as BACE 2 inhibitors which is used in the treatment of diabetes. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl picolinate is used in the preparation of 2-Aminodihydro[1,3]thiazines as BACE 2 inhibitors which is used in the treatment of diabetes. It is also used as pharmaceutical intermediate.

Solubility
Fully miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Ethyl picolinate
1°C to 3°C
239°C to 241°C
Characteristic
1.5105
100 g
ethyl picolinate, ethyl 2-picolinate, ethyl 2-pyridinecarboxylate, picolinic acid, ethyl ester, 2-picolinic acid ethyl ester, 2-pyridinecarboxylic acid, ethyl ester, 2-ethoxycarbonyl pyridine, picolinic acid ethyl ester, 2-pyridinecarboxylic acid ethyl ester, pyridine-2-carboxylic acid ethyl ester
FQYYIPZPELSLDK-UHFFFAOYSA-N
ethyl pyridine-2-carboxylate
17307
99%
2524-52-9
1.12
107°C (224°F)
C8H9NO2
MFCD00006292
122485
Fully miscible in water.
CCOC(=O)C1=CC=CC=N1
151.17
151.17

Chemical Identifiers

2524-52-9
151.17
FQYYIPZPELSLDK-UHFFFAOYSA-N
17307
CCOC(=O)C1=CC=CC=N1
C8H9NO2
MFCD00006292
ethyl picolinate, ethyl 2-picolinate, ethyl 2-pyridinecarboxylate, picolinic acid, ethyl ester, 2-picolinic acid ethyl ester, 2-pyridinecarboxylic acid, ethyl ester, 2-ethoxycarbonyl pyridine, picolinic acid ethyl ester, 2-pyridinecarboxylic acid ethyl ester, pyridine-2-carboxylic acid ethyl ester
ethyl pyridine-2-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P305+P351+P338
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

EINECSNumber : 219-758-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only