missing translation for 'onlineSavingsMsg'
Learn More

Ethyl picolinate, 99%

Catalog No. AAA1397314
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1397314 25 g
AAA1397318 50 g
AAA1397322 100 g
AAA1397330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1397314 Supplier Thermo Scientific Chemicals Supplier No. A1397314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2524-52-9 | C8H9NO2 | 151.17 g/mol

Ethyl picolinate is used in the preparation of 2-Aminodihydro[1,3]thiazines as BACE 2 inhibitors which is used in the treatment of diabetes. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl picolinate is used in the preparation of 2-Aminodihydro[1,3]thiazines as BACE 2 inhibitors which is used in the treatment of diabetes. It is also used as pharmaceutical intermediate.

Solubility
Fully miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2524-52-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00006292
InChI Key FQYYIPZPELSLDK-UHFFFAOYSA-N
Synonym ethyl picolinate, ethyl 2-picolinate, ethyl 2-pyridinecarboxylate, picolinic acid, ethyl ester, 2-picolinic acid ethyl ester, 2-pyridinecarboxylic acid, ethyl ester, 2-ethoxycarbonyl pyridine, picolinic acid ethyl ester, 2-pyridinecarboxylic acid ethyl ester, pyridine-2-carboxylic acid ethyl ester
PubChem CID 17307
IUPAC Name ethyl pyridine-2-carboxylate
SMILES CCOC(=O)C1=CC=CC=N1

Specifications

Melting Point 1°C to 3°C
Density 1.12
Boiling Point 239°C to 241°C
Flash Point 107°C (224°F)
Odor Characteristic
Refractive Index 1.5105
Quantity 25 g
Beilstein 122485
Solubility Information Fully miscible in water.
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material Ethyl picolinate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P305+P351+P338
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 219-758-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only