Learn More
Ethyl phosphorodichloridite, 98%
CAS: 1498-42-6 | C2H5Cl2OP | 146.93 g/mol
$148.64 - $2233.11
Chemical Identifiers
| CAS | 1498-42-6 |
|---|---|
| Molecular Formula | C2H5Cl2OP |
| Molecular Weight (g/mol) | 146.93 |
| MDL Number | MFCD00000525 |
| InChI Key | PVCINRPAXRJLEP-UHFFFAOYSA-N |
| Synonym | ethyl dichlorophosphite, ethyl phosphorodichloridite, ethyl dichlorophosphinite, ethylphosphorodichloridite, ethyl phosphorodichloridoite, acmc-20akws, dichloroethoxyphosphine, dichloro ethoxy phosphane, dichloro ethoxy phosphine, c2h5opcl2 |
| PubChem CID | 73905 |
| IUPAC Name | dichloro(ethoxy)phosphane |
| SMILES | CCOP(Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2117909
|
Thermo Scientific Chemicals
B2117909 |
10 g |
Each for $148.64
|
|
|||||
|
AAB2117918
|
Thermo Scientific Chemicals
B2117918 |
50 g |
Each for $487.22
|
|
|||||
|
AAB2117930
|
Thermo Scientific Chemicals
B2117930 |
250 g |
Each for $2,233.11
|
|
|||||
Description
Ethyl dichlorophosphite is used in the synthesis of peptide derivatives. It is used as an anhydride forming reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl dichlorophosphite is used in the synthesis of peptide derivatives. It is used as an anhydride forming reagent.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/moisture and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
| 1498-42-6 | |
| 146.93 | |
| PVCINRPAXRJLEP-UHFFFAOYSA-N | |
| 73905 | |
| CCOP(Cl)Cl |
| C2H5Cl2OP | |
| MFCD00000525 | |
| ethyl dichlorophosphite, ethyl phosphorodichloridite, ethyl dichlorophosphinite, ethylphosphorodichloridite, ethyl phosphorodichloridoite, acmc-20akws, dichloroethoxyphosphine, dichloro ethoxy phosphane, dichloro ethoxy phosphine, c2h5opcl2 | |
| dichloro(ethoxy)phosphane |
Specifications
| 1498-42-6 | |
| 117°C to 118°C | |
| Acrid | |
| 1.466 | |
| 10 g | |
| 1697483 | |
| ethyl dichlorophosphite, ethyl phosphorodichloridite, ethyl dichlorophosphinite, ethylphosphorodichloridite, ethyl phosphorodichloridoite, acmc-20akws, dichloroethoxyphosphine, dichloro ethoxy phosphane, dichloro ethoxy phosphine, c2h5opcl2 | |
| PVCINRPAXRJLEP-UHFFFAOYSA-N | |
| dichloro(ethoxy)phosphane | |
| 73905 | |
| 98% |
| 1.284 | |
| 4°C (39°F) | |
| C2H5Cl2OP | |
| MFCD00000525 | |
| UN2924 | |
| Moisture sensitive | |
| Reacts with water. | |
| CCOP(Cl)Cl | |
| 146.93 | |
| 146.94 | |
| Ethyl phosphorodichloridite |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber : 216-098-5
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only