accessibility menu, dialog, popup

UserName

CH3OPClCl

CAS RN 1498-42-6

IUPAC Name: ethyl dichlorophosphinite
Synonyms: ethyl dichlorophosphinite
Molecular Weight (g/mol): 146.93
Molecular Formula: C2H5Cl2OP
InChi Key: PVCINRPAXRJLEP-UHFFFAOYSA-N
SMILES: CCOP(Cl)Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)