missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl phosphorodichloridite, 98%
Description
Ethyl dichlorophosphite is used in the synthesis of peptide derivatives. It is used as an anhydride forming reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl dichlorophosphite is used in the synthesis of peptide derivatives. It is used as an anhydride forming reagent.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/moisture and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Ethyl dichlorophosphite is used in the synthesis of peptide derivatives. It is used as an anhydride forming reagent.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/moisture and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Specifications
| Density | 1.284 |
| Boiling Point | 117°C to 118°C |
| Flash Point | 4°C (39°F) |
| Odor | Acrid |
| Refractive Index | 1.466 |
| Quantity | 10 g |
| UN Number | UN2924 |
| Beilstein | 1697483 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Reacts with water. |
| Show More |
Safety and Handling
| Hazard Category | H225-H314-H335 |
| Hazard Statement | GHS H Statement H225-H314-H318 Highly flammable liquid and vapour. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
| EINECSNumber | 216-098-5 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only