missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Phosphate (Mono- and Di- Ester mixture), TCI America™

Catalog No. E023625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E023625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E023625ML Supplier TCI America Supplier No. E023625ML
Only null left

Chemical Identifiers

CAS 37203-76-2
MDL Number MFCD00136002
Synonym Phosphoric Acid Ethyl Ester

Specifications

Color Yellow
Quantity 25 mL
UN Number 1760
Physical Form Liquid
Chemical Name or Material Ethyl Phosphate (Mono- and Di- Ester mixture)
EINECSNumber (2)-1986
TSCA Yes