accessibility menu, dialog, popup

UserName

H3COHOHPOHOHO

CAS RN 37203-76-2

IUPAC Name: ethanol; phosphoric acid
Synonyms: ethyl alcohol; phosphoric acid
Molecular Weight (g/mol): 144.06
Molecular Formula: C2H9O5P
InChi Key: PEZBJHXXIFFJBI-UHFFFAOYSA-N
SMILES: CCO.OP(O)(O)=O
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results