missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (Phenylthio)acetate 97.0+%, TCI America™
$57.79 - $150.98
Chemical Identifiers
| CAS | 7605-25-6 |
|---|---|
| Molecular Formula | C10H12O2S |
| Molecular Weight (g/mol) | 196.264 |
| MDL Number | MFCD00145107 |
| InChI Key | SEDRTXNDGKRHBL-UHFFFAOYSA-N |
| Synonym | ethyl phenylthio acetate, ethyl 2-phenylthio acetate, ethyl 2-phenylsulfanyl acetate, acetic acid, phenylthio-, ethyl ester, ethyl 2-phenylthioacetate, phenylthio acetic acid ethyl ester, phenylthio acetic acid, ethyl ester, ethyl phenylsulfanyl acetate, pubchem10853 |
| PubChem CID | 97312 |
| IUPAC Name | ethyl 2-phenylsulfanylacetate |
| SMILES | CCOC(=O)CSC1=CC=CC=C1 |
Chemical Identifiers
| 7605-25-6 | |
| 196.264 | |
| SEDRTXNDGKRHBL-UHFFFAOYSA-N | |
| 97312 | |
| CCOC(=O)CSC1=CC=CC=C1 |
| C10H12O2S | |
| MFCD00145107 | |
| ethyl phenylthio acetate, ethyl 2-phenylthio acetate, ethyl 2-phenylsulfanyl acetate, acetic acid, phenylthio-, ethyl ester, ethyl 2-phenylthioacetate, phenylthio acetic acid ethyl ester, phenylthio acetic acid, ethyl ester, ethyl phenylsulfanyl acetate, pubchem10853 | |
| ethyl 2-phenylsulfanylacetate |
Specifications
| 7605-25-6 | |
| 160°C | |
| MFCD00145107 | |
| ethyl phenylthio acetate, ethyl 2-phenylthio acetate, ethyl 2-phenylsulfanyl acetate, acetic acid, phenylthio-, ethyl ester, ethyl 2-phenylthioacetate, phenylthio acetic acid ethyl ester, phenylthio acetic acid, ethyl ester, ethyl phenylsulfanyl acetate, pubchem10853 | |
| CCOC(=O)CSC1=CC=CC=C1 | |
| 196.264 | |
| 196.26 | |
| Liquid |
| Yellow | |
| C10H12O2S | |
| 5 g | |
| SEDRTXNDGKRHBL-UHFFFAOYSA-N | |
| ethyl 2-phenylsulfanylacetate | |
| 97312 | |
| ≥97.0% (GC) | |
| Ethyl (Phenylthio)acetate |
Safety and Handling
TSCA : No