missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Phenylsulfonylacetate 98.0+%, TCI America™

Catalog No. P13065G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13065G 5 g
P130625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13065G Supplier TCI America Supplier No. P13065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7605-30-3
Molecular Formula C10H12O4S
Molecular Weight (g/mol) 228.262
MDL Number MFCD00025041
InChI Key NJBWORPRIRNTLH-UHFFFAOYSA-N
Synonym Phenylsulfonylacetic Acid Ethyl Ester
PubChem CID 82078
IUPAC Name ethyl 2-(benzenesulfonyl)acetate
SMILES CCOC(=O)CS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 44°C
Color White
Quantity 5 g
Formula Weight 228.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Phenylsulfonylacetate