missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Phenylpropiolate 97.0+%, TCI America™
Supplier: TCI America P081425G
Specifications
| Ethyl Phenylpropiolate | |
| Yellow | |
| C11H10O2 | |
| 25 g | |
| ACJOYTKWHPEIHW-UHFFFAOYSA-N | |
| ethyl 3-phenylprop-2-ynoate | |
| 91516 | |
| ≥97.0% (GC) |
| 2216-94-6 | |
| 103°C | |
| MFCD00009185 | |
| ethyl phenylpropiolate, ethylphenylpropiolate, ethyl 3-phenylpropiolate, ethyl phenylpropriolate, 2-propynoic acid, 3-phenyl-, ethyl ester, ethyl 3-phenylpropynoate, ethyl phenylpropynoate, ethyl phenylacetylenecarboxylate, phenylpropiolic acid ethyl ester, ethyl 3-phenyl-2-propynoate | |
| CCOC(=O)C#CC1=CC=CC=C1 | |
| 174.199 | |
| 174.20 | |
| Liquid |
Chemical Identifiers
| 2216-94-6 | |
| 174.199 | |
| ACJOYTKWHPEIHW-UHFFFAOYSA-N | |
| 91516 | |
| CCOC(=O)C#CC1=CC=CC=C1 |
| C11H10O2 | |
| MFCD00009185 | |
| ethyl phenylpropiolate, ethylphenylpropiolate, ethyl 3-phenylpropiolate, ethyl phenylpropriolate, 2-propynoic acid, 3-phenyl-, ethyl ester, ethyl 3-phenylpropynoate, ethyl phenylpropynoate, ethyl phenylacetylenecarboxylate, phenylpropiolic acid ethyl ester, ethyl 3-phenyl-2-propynoate | |
| ethyl 3-phenylprop-2-ynoate |
Safety and Handling
RTECSNumber : UE0110000
TSCA : No
Recommended Storage : Refrigerator