accessibility menu, dialog, popup

UserName

CAS RN 2216-94-6

IUPAC Name: ethyl 3-phenylprop-2-ynoate
Synonyms: ethyl 3-phenylprop-2-ynoate
Molecular Weight (g/mol): 174.2
Molecular Formula: C11H10O2
InChi Key: ACJOYTKWHPEIHW-UHFFFAOYSA-N
SMILES: CCOC(=O)C#CC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)