missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl phenylpropiolate, 98%
CAS: 2216-94-6 | C11H10O2 | 174.2 g/mol
$241.68 - $241.68
Chemical Identifiers
| CAS | 2216-94-6 |
|---|---|
| Molecular Formula | C11H10O2 |
| Molecular Weight (g/mol) | 174.2 |
| MDL Number | MFCD00009185 |
| InChI Key | ACJOYTKWHPEIHW-UHFFFAOYSA-N |
| Synonym | ethyl phenylpropiolate, ethylphenylpropiolate, ethyl 3-phenylpropiolate, ethyl phenylpropriolate, 2-propynoic acid, 3-phenyl-, ethyl ester, ethyl 3-phenylpropynoate, ethyl phenylpropynoate, ethyl phenylacetylenecarboxylate, phenylpropiolic acid ethyl ester, ethyl 3-phenyl-2-propynoate |
| PubChem CID | 91516 |
| IUPAC Name | ethyl 3-phenylprop-2-ynoate |
| SMILES | CCOC(=O)C#CC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC118850250
|
Thermo Scientific Chemicals
118850250 |
25 g | Glass bottle |
Each for $241.68
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2216-94-6 | |
| 174.2 | |
| ACJOYTKWHPEIHW-UHFFFAOYSA-N | |
| 91516 | |
| CCOC(=O)C#CC1=CC=CC=C1 |
| C11H10O2 | |
| MFCD00009185 | |
| ethyl phenylpropiolate, ethylphenylpropiolate, ethyl 3-phenylpropiolate, ethyl phenylpropriolate, 2-propynoic acid, 3-phenyl-, ethyl ester, ethyl 3-phenylpropynoate, ethyl phenylpropynoate, ethyl phenylacetylenecarboxylate, phenylpropiolic acid ethyl ester, ethyl 3-phenyl-2-propynoate | |
| ethyl 3-phenylprop-2-ynoate |
Specifications
| 2216-94-6 | |
| 1.0500g/mL | |
| >112°C | |
| 98% | |
| C11H10O2 | |
| C6H5C∽CCO2C2H5 | |
| 25 g | |
| 1.05 | |
| ACJOYTKWHPEIHW-UHFFFAOYSA-N | |
| ethyl 3-phenylprop-2-ynoate | |
| 91516 | |
| 98% | |
| Ethyl phenylpropiolate, 98% |
| Yellow | |
| 260°C to 270°C | |
| Authentic | |
| Glass bottle | |
| 1.5510 to 1.553 | |
| MFCD00009185 | |
| 09, 634 | |
| ethyl phenylpropiolate, ethylphenylpropiolate, ethyl 3-phenylpropiolate, ethyl phenylpropriolate, 2-propynoic acid, 3-phenyl-, ethyl ester, ethyl 3-phenylpropynoate, ethyl phenylpropynoate, ethyl phenylacetylenecarboxylate, phenylpropiolic acid ethyl ester, ethyl 3-phenyl-2-propynoate | |
| CCOC(=O)C#CC1=CC=CC=C1 | |
| 174.2 | |
| 174.2 | |
| Liquid |
Safety and Handling
EINECSNumber : 218-703-8
RTECSNumber : UE0110000
RUO – Research Use Only