missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenylpropiolate, 98+%

Catalog No. AAL0060306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0060306 5 g
AAL0060314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0060306 Supplier Thermo Scientific Chemicals Supplier No. L0060306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2216-94-6 | C11H10O2 | 174.199 g/mol

Ethyl phenylpropiolate, is an important organic intermediate. It is also used as an intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl phenylpropiolate, is an important organic intermediate. It is also used as an intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Heat sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 2216-94-6
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.199
MDL Number MFCD00009185
InChI Key ACJOYTKWHPEIHW-UHFFFAOYSA-N
Synonym ethyl phenylpropiolate, ethylphenylpropiolate, ethyl 3-phenylpropiolate, ethyl phenylpropriolate, 2-propynoic acid, 3-phenyl-, ethyl ester, ethyl 3-phenylpropynoate, ethyl phenylpropynoate, ethyl phenylacetylenecarboxylate, phenylpropiolic acid ethyl ester, ethyl 3-phenyl-2-propynoate
PubChem CID 91516
IUPAC Name ethyl 3-phenylprop-2-ynoate
SMILES CCOC(=O)C#CC1=CC=CC=C1

Specifications

Density 1.055
Boiling Point 152°C to 153°C (21 mmHg)
Flash Point >110°C (230°F)
Odor Odorless
Refractive Index 1.552
Quantity 5 g
Beilstein 639637
Solubility Information Insoluble in water.
Formula Weight 174.2
Percent Purity ≥98%
Chemical Name or Material Ethyl phenylpropiolate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 218-703-8
RTECSNumber UE0110000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only