Learn More
Ethyl phenylglyoxylate, 98%
CAS: 1603-79-8 | C10H10O3 | 178.19 g/mol
Supplier: Thermo Scientific Chemicals B2529214
Description
Ethyl phenylglyoxylate has been used in preparation of 1,5-dihydro-5-deazaflavin derivatives possessing a chiral substituent at N(3) position.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl phenylglyoxylate has been used in preparation of 1,5-dihydro-5-deazaflavin derivatives possessing a chiral substituent at N(3) position.
Solubility
Soluble in water 1143 mg/L @ 25°C .
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Specifications
| Ethyl phenylglyoxylate | |
| 1.122 | |
| >110°C (230°F) | |
| 1.516 | |
| 25 g | |
| ethyl benzoylformate, ethyl phenylglyoxylate, ethyl oxophenylacetate, ethylbenzoylformate, phenylglyoxylic acid ethyl ester, ethyl oxo phenyl acetate, benzeneacetic acid, .alpha.-oxo-, ethyl ester, benzoylformic acid ethyl ester, ethyl benzoyl formate, glyoxylic acid, phenyl-, ethyl ester | |
| QKLCQKPAECHXCQ-UHFFFAOYSA-N | |
| ethyl 2-oxo-2-phenylacetate | |
| 15349 | |
| 178.19 |
| 1603-79-8 | |
| 138°C to 139°C (18 mmHg) | |
| C10H10O3 | |
| MFCD00009120 | |
| Moisture sensitive | |
| Soluble in water 1143mg/L @ 25°C . | |
| CCOC(=O)C(=O)C1=CC=CC=C1 | |
| 178.19 | |
| CHEBI:84260 | |
| 98% |
Chemical Identifiers
| 1603-79-8 | |
| 178.19 | |
| QKLCQKPAECHXCQ-UHFFFAOYSA-N | |
| 15349 | |
| ethyl 2-oxo-2-phenylacetate |
| C10H10O3 | |
| MFCD00009120 | |
| ethyl benzoylformate, ethyl phenylglyoxylate, ethyl oxophenylacetate, ethylbenzoylformate, phenylglyoxylic acid ethyl ester, ethyl oxo phenyl acetate, benzeneacetic acid, .alpha.-oxo-, ethyl ester, benzoylformic acid ethyl ester, ethyl benzoyl formate, glyoxylic acid, phenyl-, ethyl ester | |
| CHEBI:84260 | |
| CCOC(=O)C(=O)C1=CC=CC=C1 |
Safety and Handling
EINECSNumber : 216-504-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only