missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Phenylcyanoacetate 95.0+%, TCI America™
$94.33 - $377.33
Chemical Identifiers
| CAS | 4553-07-5 |
|---|---|
| Molecular Formula | C11H11NO2 |
| Molecular Weight (g/mol) | 189.21 |
| MDL Number | MFCD00001866 |
| InChI Key | SXIRJEDGTAKGKU-UHFFFAOYNA-N |
| Synonym | ethyl phenylcyanoacetate, phenylcyanoacetic acid ethyl ester, ethyl cyanophenylacetate, ethyl cyano phenyl acetate, ethyl alpha-cyanophenylacetate, ethyl alpha-cyano-alpha-phenylacetate, acetic acid, cyanophenyl-, ethyl ester, alpha-cyanobenzeneacetic acid, ethyl ester, benzeneacetic acid, alpha-cyano-, ethyl ester, benzeneacetic acid, .alpha.-cyano-, ethyl ester |
| PubChem CID | 95298 |
| ChEBI | CHEBI:51926 |
| IUPAC Name | ethyl 2-cyano-2-phenylacetate |
| SMILES | CCOC(=O)C(C#N)C1=CC=CC=C1 |
Chemical Identifiers
| 4553-07-5 | |
| 189.21 | |
| SXIRJEDGTAKGKU-UHFFFAOYNA-N | |
| 95298 | |
| ethyl 2-cyano-2-phenylacetate |
| C11H11NO2 | |
| MFCD00001866 | |
| ethyl phenylcyanoacetate, phenylcyanoacetic acid ethyl ester, ethyl cyanophenylacetate, ethyl cyano phenyl acetate, ethyl alpha-cyanophenylacetate, ethyl alpha-cyano-alpha-phenylacetate, acetic acid, cyanophenyl-, ethyl ester, alpha-cyanobenzeneacetic acid, ethyl ester, benzeneacetic acid, alpha-cyano-, ethyl ester, benzeneacetic acid, .alpha.-cyano-, ethyl ester | |
| CHEBI:51926 | |
| CCOC(=O)C(C#N)C1=CC=CC=C1 |
Specifications
| 4553-07-5 | |
| 275°C | |
| MFCD00001866 | |
| ethyl phenylcyanoacetate, phenylcyanoacetic acid ethyl ester, ethyl cyanophenylacetate, ethyl cyano phenyl acetate, ethyl alpha-cyanophenylacetate, ethyl alpha-cyano-alpha-phenylacetate, acetic acid, cyanophenyl-, ethyl ester, alpha-cyanobenzeneacetic acid, ethyl ester, benzeneacetic acid, alpha-cyano-, ethyl ester, benzeneacetic acid, .alpha.-cyano-, ethyl ester | |
| CCOC(=O)C(C#N)C1=CC=CC=C1 | |
| 189.21 | |
| CHEBI:51926 | |
| ≥95.0% (GC) | |
| Ethyl Phenylcyanoacetate |
| Yellow | |
| C11H11NO2 | |
| 5 g | |
| SXIRJEDGTAKGKU-UHFFFAOYNA-N | |
| ethyl 2-cyano-2-phenylacetate | |
| 95298 | |
| 189.21 | |
| Liquid |
Safety and Handling
TSCA : No