accessibility menu, dialog, popup

UserName

H3COON

CAS RN 4553-07-5

IUPAC Name: ethyl 2-cyano-2-phenylacetate
Synonyms: ethyl phenylcyanoacetate
Molecular Weight (g/mol): 189.21
Molecular Formula: C11H11NO2
InChi Key: SXIRJEDGTAKGKU-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C#N)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results