missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Phenylcyanoacetate 95.0+%, TCI America™

Catalog No. P12555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12555G 5 g
P125525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12555G Supplier TCI America Supplier No. P12555G

Chemical Identifiers

CAS 4553-07-5
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00001866
InChI Key SXIRJEDGTAKGKU-UHFFFAOYNA-N
Synonym ethyl phenylcyanoacetate, phenylcyanoacetic acid ethyl ester, ethyl cyanophenylacetate, ethyl cyano phenyl acetate, ethyl alpha-cyanophenylacetate, ethyl alpha-cyano-alpha-phenylacetate, acetic acid, cyanophenyl-, ethyl ester, alpha-cyanobenzeneacetic acid, ethyl ester, benzeneacetic acid, alpha-cyano-, ethyl ester, benzeneacetic acid, .alpha.-cyano-, ethyl ester
PubChem CID 95298
ChEBI CHEBI:51926
IUPAC Name ethyl 2-cyano-2-phenylacetate
SMILES CCOC(=O)C(C#N)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 275°C
Quantity 5 g
Formula Weight 189.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Phenylcyanoacetate