missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenylcyanoacetate, 98%

CAS: 4553-07-5 | C11H11NO2 | 189.21 g/mol

$83.68 - $293.04

Chemical Identifiers

CAS 4553-07-5
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00001866
InChI Key SXIRJEDGTAKGKU-UHFFFAOYNA-N
Synonym ethyl phenylcyanoacetate, phenylcyanoacetic acid ethyl ester, ethyl cyanophenylacetate, ethyl cyano phenyl acetate, ethyl alpha-cyanophenylacetate, ethyl alpha-cyano-alpha-phenylacetate, acetic acid, cyanophenyl-, ethyl ester, alpha-cyanobenzeneacetic acid, ethyl ester, benzeneacetic acid, alpha-cyano-, ethyl ester, benzeneacetic acid, .alpha.-cyano-, ethyl ester
PubChem CID 95298
ChEBI CHEBI:51926
IUPAC Name ethyl 2-cyano-2-phenylacetate
SMILES CCOC(=O)C(C#N)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0392906
View Documents
Thermo Scientific Chemicals
L0392906
5 g
Each for $83.68
Only null left
 
AAL0392914
View Documents
Thermo Scientific Chemicals
L0392914
25 g
Each for $293.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4553-07-5
189.21
SXIRJEDGTAKGKU-UHFFFAOYNA-N
95298
ethyl 2-cyano-2-phenylacetate
C11H11NO2
MFCD00001866
ethyl phenylcyanoacetate, phenylcyanoacetic acid ethyl ester, ethyl cyanophenylacetate, ethyl cyano phenyl acetate, ethyl alpha-cyanophenylacetate, ethyl alpha-cyano-alpha-phenylacetate, acetic acid, cyanophenyl-, ethyl ester, alpha-cyanobenzeneacetic acid, ethyl ester, benzeneacetic acid, alpha-cyano-, ethyl ester, benzeneacetic acid, .alpha.-cyano-, ethyl ester
CHEBI:51926
CCOC(=O)C(C#N)C1=CC=CC=C1

Specifications

4553-07-5
274°C to 275°C
C11H11NO2
MFCD00001866
611232
SXIRJEDGTAKGKU-UHFFFAOYNA-N
ethyl 2-cyano-2-phenylacetate
95298
189.21
Ethyl phenylcyanoacetate
1.09
>110°C (230°F)
1.505
5 g
ethyl phenylcyanoacetate, phenylcyanoacetic acid ethyl ester, ethyl cyanophenylacetate, ethyl cyano phenyl acetate, ethyl alpha-cyanophenylacetate, ethyl alpha-cyano-alpha-phenylacetate, acetic acid, cyanophenyl-, ethyl ester, alpha-cyanobenzeneacetic acid, ethyl ester, benzeneacetic acid, alpha-cyano-, ethyl ester, benzeneacetic acid, .alpha.-cyano-, ethyl ester
CCOC(=O)C(C#N)C1=CC=CC=C1
189.21
CHEBI:51926
98%
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 224-921-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only