missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Phenyl Sulfide 98.0+%, TCI America™

Catalog No. E042610G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E042610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E042610G Supplier TCI America Supplier No. E042610G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 622-38-8
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.23
MDL Number MFCD00009265
InChI Key AEHWKBXBXYNPCX-UHFFFAOYSA-N
Synonym ethyl phenyl sulfide, ethylthio benzene, benzene, ethylthio, thiophenetole, phenyl ethyl sulfide, ethylphenylsulfide, phenylthio ethane, ethylsulfanyl benzene, sulfide, ethyl phenyl, 1-thiapropyl benzene
PubChem CID 12144
IUPAC Name (ethylsulfanyl)benzene
SMILES CCSC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 205°C
Quantity 10 g
Formula Weight 138.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Phenyl Sulfide