Learn More
Ethyl phenyl sulfide, 98%
CAS: 622-38-8 | C8H10S | 138.23 g/mol
Supplier: Thermo Scientific Chemicals A1398714
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ethyl phenyl sulfide | |
| 1.02 | |
| 73°C (163°F) | |
| C8H10S | |
| MFCD00009265 | |
| 1905731 | |
| AEHWKBXBXYNPCX-UHFFFAOYSA-N | |
| ethylsulfanylbenzene | |
| 12144 | |
| 98% |
| 622-38-8 | |
| 204°C to 205°C | |
| Stench | |
| 1.5655 | |
| 25 g | |
| ethyl phenyl sulfide, ethylthio benzene, benzene, ethylthio, thiophenetole, phenyl ethyl sulfide, ethylphenylsulfide, phenylthio ethane, ethylsulfanyl benzene, sulfide, ethyl phenyl, 1-thiapropyl benzene | |
| CCSC1=CC=CC=C1 | |
| 138.23 | |
| 138.23 |
Chemical Identifiers
| 622-38-8 | |
| 138.23 | |
| AEHWKBXBXYNPCX-UHFFFAOYSA-N | |
| 12144 | |
| CCSC1=CC=CC=C1 |
| C8H10S | |
| MFCD00009265 | |
| ethyl phenyl sulfide, ethylthio benzene, benzene, ethylthio, thiophenetole, phenyl ethyl sulfide, ethylphenylsulfide, phenylthio ethane, ethylsulfanyl benzene, sulfide, ethyl phenyl, 1-thiapropyl benzene | |
| ethylsulfanylbenzene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 210-731-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only