missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenyl sulfide, 98%

Catalog No. AAA1398722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1398722 100 g
AAA1398706 5 g
AAA1398714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1398722 Supplier Thermo Scientific Chemicals Supplier No. A1398722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 622-38-8 | C8H10S | 138.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 622-38-8
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.23
MDL Number MFCD00009265
InChI Key AEHWKBXBXYNPCX-UHFFFAOYSA-N
Synonym ethyl phenyl sulfide, ethylthio benzene, benzene, ethylthio, thiophenetole, phenyl ethyl sulfide, ethylphenylsulfide, phenylthio ethane, ethylsulfanyl benzene, sulfide, ethyl phenyl, 1-thiapropyl benzene
PubChem CID 12144
IUPAC Name ethylsulfanylbenzene
SMILES CCSC1=CC=CC=C1

Specifications

Density 1.02
Boiling Point 204°C to 205°C
Flash Point 73°C (163°F)
Odor Stench
Refractive Index 1.5655
Quantity 100 g
Beilstein 1905731
Formula Weight 138.23
Percent Purity 98%
Chemical Name or Material Ethyl phenyl sulfide
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 210-731-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only