missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenoxyacetate, 99%

Catalog No. AAA1415609
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1415609 10 g
AAA1415618 50 g
AAA1415630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1415609 Supplier Thermo Scientific Chemicals Supplier No. A1415609

CAS: 2555-49-9 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2555-49-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00026895
InChI Key MGZFVSUXQXCEHM-UHFFFAOYSA-N
Synonym ethyl phenoxyacetate, acetic acid, phenoxy-, ethyl ester, phenoxyacetic acid ethyl ester, acetic acid, 2-phenoxy-, ethyl ester, phenoxy-acetic acid ethyl ester, phenoxyacetic acid, ethyl ester, ethyl2-phenoxyacetate, pubchem12485, acmc-1ckja, ethylphenoxyacetate
PubChem CID 17365
IUPAC Name ethyl 2-phenoxyacetate
SMILES CCOC(=O)COC1=CC=CC=C1

Specifications

Density 1.108
Boiling Point 251°C to 253°C
Flash Point >110°C (230°F)
Refractive Index 1.5055
Quantity 10 g
Beilstein 1871712
Formula Weight 180.2
Percent Purity 99%
Chemical Name or Material Ethyl phenoxyacetate
EINECSNumber 219-867-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only