missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Pentafluorobenzoate 98.0+%, TCI America™
$50.05 - $136.69
Chemical Identifiers
| CAS | 4522-93-4 |
|---|---|
| Molecular Formula | C9H5F5O2 |
| Molecular Weight (g/mol) | 240.13 |
| MDL Number | MFCD00039211 |
| InChI Key | DFUDMSIRGGTHGI-UHFFFAOYSA-N |
| Synonym | ethyl pentafluorobenzoate, ethyl perfluorobenzoate, pentafluorobenzoic acid ethyl ester, 2,3,4,5,6-pentafluoro-benzoic acid ethyl ester, benzoic acid, pentafluoro-, ethyl ester, ethylpentafluorobenzoate, acmc-1ajj1, rarechem al bi 0017, perfluorobenzoic acid ethyl ester |
| PubChem CID | 78283 |
| IUPAC Name | ethyl 2,3,4,5,6-pentafluorobenzoate |
| SMILES | CCOC(=O)C1=C(F)C(F)=C(F)C(F)=C1F |
Chemical Identifiers
| 4522-93-4 | |
| 240.13 | |
| DFUDMSIRGGTHGI-UHFFFAOYSA-N | |
| 78283 | |
| CCOC(=O)C1=C(F)C(F)=C(F)C(F)=C1F |
| C9H5F5O2 | |
| MFCD00039211 | |
| ethyl pentafluorobenzoate, ethyl perfluorobenzoate, pentafluorobenzoic acid ethyl ester, 2,3,4,5,6-pentafluoro-benzoic acid ethyl ester, benzoic acid, pentafluoro-, ethyl ester, ethylpentafluorobenzoate, acmc-1ajj1, rarechem al bi 0017, perfluorobenzoic acid ethyl ester | |
| ethyl 2,3,4,5,6-pentafluorobenzoate |
Specifications
| 4522-93-4 | |
| 113°C | |
| MFCD00039211 | |
| ethyl pentafluorobenzoate, ethyl perfluorobenzoate, pentafluorobenzoic acid ethyl ester, 2,3,4,5,6-pentafluoro-benzoic acid ethyl ester, benzoic acid, pentafluoro-, ethyl ester, ethylpentafluorobenzoate, acmc-1ajj1, rarechem al bi 0017, perfluorobenzoic acid ethyl ester | |
| CCOC(=O)C1=C(F)C(F)=C(F)C(F)=C1F | |
| 240.13 | |
| 240.13 | |
| Liquid |
| Colorless | |
| C9H5F5O2 | |
| 5 g | |
| DFUDMSIRGGTHGI-UHFFFAOYSA-N | |
| ethyl 2,3,4,5,6-pentafluorobenzoate | |
| 78283 | |
| ≥98.0% (GC) | |
| Ethyl Pentafluorobenzoate |
Safety and Handling
TSCA : No