missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Pentafluorobenzoate 98.0+%, TCI America™

Catalog No. P14195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14195G 5 g
P141925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14195G Supplier TCI America Supplier No. P14195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4522-93-4
Molecular Formula C9H5F5O2
Molecular Weight (g/mol) 240.13
MDL Number MFCD00039211
InChI Key DFUDMSIRGGTHGI-UHFFFAOYSA-N
Synonym ethyl pentafluorobenzoate, ethyl perfluorobenzoate, pentafluorobenzoic acid ethyl ester, 2,3,4,5,6-pentafluoro-benzoic acid ethyl ester, benzoic acid, pentafluoro-, ethyl ester, ethylpentafluorobenzoate, acmc-1ajj1, rarechem al bi 0017, perfluorobenzoic acid ethyl ester
PubChem CID 78283
IUPAC Name ethyl 2,3,4,5,6-pentafluorobenzoate
SMILES CCOC(=O)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Color Colorless
Boiling Point 113°C
Quantity 5 g
Formula Weight 240.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Pentafluorobenzoate